missing translation for 'onlineSavingsMsg'
Learn More

LMX1A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA534470

Product Code. 15315401

  • 514.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

A suggested positive control is rat brain tissue lysate. PA5-34470 can be used with blocking peptide PEP-1512.

The LIM homeobox transcription factor 1 (LMX1A) belongs to the LIM-homeodomain family. Members of this family are known to be important for pattern formation during development. LMX1A binds an A/T-rich sequence and functions in the nucleus as a transcriptional activator to the insulin gene promoter. In the developing embryo, LMX1A is expressed along the neuraxis and leads to the development of the roof plate of the vertebrae. While LMX1A is crucial for the differentiation of dopamine neurons, the related LMX1B appears to be important for both the development and the survival of these cells. It has been suggested that LMX1A might be a useful biomarker or risk factor in tumor aggressiveness.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LMX1A
Polyclonal
Unconjugated
Lmx1a
dr; dreher; LIM homeobox transcription factor 1 alpha; LIM homeobox transcription factor 1, alpha; LIM homeobox transcription factor 1-alpha; LIM/homeobox protein 1.1; LIM/homeobox protein LMX1A; LMX1; LMX-1; Lmx1.1; LMX-1.1; Lmx1a; MGC87616; shaker short-tail; sst
Rabbit
Antigen affinity chromatography
RUO
110648, 289201, 4009
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8TE12, Q9JKU8
Lmx1a
A 16 amino acid peptide near the carboxy terminus of human LMX1A
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.