Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points UKAS Calibration
LO-tox™ lab thermometer complete with calibration certificate.
Brand:  Brannan™ 44/810/9 + 49/503/0
 View more versions of this product
Product Code. 17853068
  • 184.00 € / Each
Explore more special offers
Description
Description
  • LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
  • Safe non-mercury filling
  • Supplied with calibration certificate
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifications
Specifications
0°C, 50°C, 100°C
Total Immersion
Glass
230°F
-4°F
-4°F to +230°F
± 1.5°C at 90°C
1.2 cm
ISO 17025 UKAS
30.5 cm
Blue Spirit
+110°C
-20°C
-20°C to +110°C
0.5°C