Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points UKAS Calibration
GREENER_CHOICE
LO-tox™ lab thermometer complete with calibration certificate.
Brand:  Brannan™ 44/826/8 + 49/503/0
 View more versions of this product
Product Code. 17872988
  • 188.00 € / Each
Explore more special offers
Description
Description
  • LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
  • Safe non-mercury filling
  • Supplied with calibration certificate
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifications
Specifications
0°C, 100°C, 250°C
Partial Immersion
Glass
680°F
14°F
14°F to 680°F
± 2.0°C at 90°C
76 mm
ISO 17025 UKAS
30.5 cm
Brown Spirit
+360°C
-10°C
-10°C to +360°C
2°C
1.2 cm