missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Long Crucible Tongs with Corrugated Tips, 20 in.

Fisherbrand™ Crucible Tongs are made from ASTM Standard (899-94) Series 400 Stainless Steel.

Brand:  Fisherbrand™ 13820023

Product Code. 17394561

  • 288.00 € / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The long design facilitates use when reaching into extreme hot or cold environments. These tongs feature integral thumb and finger openings.

  • Long-neck serrated tips
  • Raised riveted joing
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods

Certifications

ISO 9001-02, GMP, cGMP

Specifications

Specifications

Crucibles
20 in.
508 mm
Crucible Tongs
Tongs
1
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers