Learn More
Lenz Laborglasintrumente Long Ground Sockets
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100624
 View more versions of this product
Product Code. 10346021
  • 105.00 € / Pack of 10
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Conical ground sockets
  • Made of DURAN™ tubing
Specifications
Specifications
10/Pk.
2 mm
Borosilicate Glass (3.3)
NS 24/40
28 mm