Learn More
AccuGENE™ Molecular Biology Water, Lonza™
Product Code. 11548015
Click to view available options
Quantity:
1 L
Ready-to-use for a wide range of molecular biology applications. AccuGENE™ Molecular Biology Water is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.
  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 L
For research or manufacturing use only.