missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ AccuGENE™ PBS
Ready to use solutions ideal for a wide range of molecular biology applications
64.45 € - 89.09 €
Specifications
Content And Storage | 18° to 26°C |
---|---|
Quantity | 1000mL |
For Use With (Application) | Ideal for a wide range of molecular biology applications |
pH | 7.4 |
Product Code | Brand | Dilution | Formulation | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dilution | Formulation | Price | Quantity & Availability | ||||
11578025
|
Lonza™
51225 |
1 X PBS | KH2PO4 1.7mM, Na2HPO4 5mM, NaCl 150mM | N/A | |||||
11588025
|
Lonza™
51226 |
10 X PBS | KH2PO4 0.017M, Na2HPO4 0.05M, NaCl 1.5M |
89.09 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
AccuGENE Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications. Each reagent is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.
- Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency.
- High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted.
- Efficient - Ready-made solutions eliminate experiment preparation time.
- Flexible - Customized solutions are available to meet individual needs
Specifications
18° to 26°C | |
1000mL | |
Ideal for a wide range of molecular biology applications | |
7.4 |