Learn More
Lonza™ IsoGel™ IEF Plates
Precast agarose IEF plates provide rapid and reproducible isoelectric focusing of proteins up to 2000kDa
Brand:  Lonza™ 56024
 View more versions of this product
Product Code. 12890125
  • 877.00 € / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Specifically designed for antibody analysis of
  • Each gel is supported on GelBond™ film for easy handling and dimensional stability
  • Gel size is compatible with most IEF chambers
  • 125 x 100mm
Isoelectric focusing, antibody separation and analysis
Order Info
Accessories packs (Cat. No. BMA56014 and BMA56010) and blotting paper (BMA56007) available separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7 to 11
Isoelectric Focusing
Isoelectric focusing, Antibody separation and analysis
Gel Plate
Solid