Learn More
Lonza™ Latitude™ HT Precast Gels
Ready-to-use gels for high-throughput DNA electrophoresis. Latitude™ HT Precast Gels are precision cast for accurate, reproducible resolution of DNA fragments for 8bp to 10kb with our SeaKem™ or NuSieve™ Agarose.
Brand:  Lonza™ 57234
 View more versions of this product
Product Code. 11517885
  • 589.09 € / Each
Explore more special offers
Alert:
Ethidium bromide is a powerful mutagen; gloves should be worn when opening pouches and handling gels and any solutions containing ethidium bromide
Description
Description
  • Provide rapid, reproducible DNA resolution of 8bp to ≥10kb
  • Manufactured from high-quality molecular biology grade SeaKem™ and NuSieve™ Brand Agarose
  • Quality and performance tested
  • Fit most large submerged electrophoresis chambers
  • Compatible with 8- and 12-tip multichannel pipets and microtiter plate spacing
  • Precision cast in accordance with ISO 9001 standards
  • Come in 100- and 200-well formats
  • Prestained with 0.5μg/mL of ethidium bromide
  • Permit runs to be completed in 30 minutes
  • Allow up to 200 samples to be run on a single precast gel
†Use of the reusable TruBand Anchor (sold separately) is required to ensure optimal gel performance
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TBE
Precast Gel
4 x 50-well
1% SeaKem™ LE Plus Agarose, 1X TBE, prestained with 0.5μg/mL of Ethidium Bromide
Agarose
Yes
0.25 in.
10 to 12 μL
14 cm
1%
300 to ≥8000 bp
Yes
High throughput DNA electrophoresis
5 Gels
18° to 26°C
0.65 cm
24 cm