missing translation for 'onlineSavingsMsg'
Learn More

Lopinavir, 98%, Thermo Scientific Chemicals

Product Code. 15315088 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15315088

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lopinavir, CAS # 192725-17-0, is an antiretroviral drug of the amphetamine class.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Lopinavir is a retroviral protease inhibitor that acts as an antiretroviral and can prevent the propagation of human immunodeficiency virus (HIV) and hepatitis viruses within cells

• Lopinavir is a dicarboxylic acid diamide and a substituted form of amphetamine

Applications

• Lopinavir may be used in the laboratory study of other retroviruses

• Lopinavir is suitable for laboratory research of drug delivery systems; some studies have shown that the activity of lopinavir can be enhanced by alternative delivery methods such as microencapsulation

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192725-17-0
Molecular Formula C37H48N4O5
Molecular Weight (g/mol) 628.81
MDL Number MFCD22628840
InChI Key KJHKTHWMRKYKJE-SUGCFTRWSA-N
PubChem CID 133109001
IUPAC Name N-[(2R,4R,5R)-5-[[2-(2,6-dimethylphenoxy)acetyl]amino]-4-hydroxy-1,6-diphenylhexan-2-yl]-3-methyl-2-(2-oxo-1,3-diazinan-1-yl)butanamide
SMILES CC(C)[C@H](N1CCCNC1=O)C(=O)N[C@H](C[C@H](O)[C@H](CC1=CC=CC=C1)NC(=O)COC1=C(C)C=CC=C1C)CC1=CC=CC=C1

Specifications

Quantity 1 g
Chemical Name or Material Lopinavir

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.