Learn More
Lovibond™ COD Tube Tests; MR; Mercury Free
Premeasured unit dose reagent for water quality testing
Brand:  Lovibond™ 2420711
 View more versions of this product
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery. Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15322055
  • 81.80 € / Pack of 25
Explore more special offers
Description
Description
Premeasured unit dose reagent for water quality testing
  • For use in SC400™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed vials eliminate risk of cross-contamination
  • Extended shelf life of up to five years
For Municipal Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted test method
Specifications
Specifications
US EPA
SC400 colorimeters and Aqua comparator™ test kits, Hach™ instruments
0 to 1500mg/L
Municipal wastewater testing, industrial wastewater and process control testing, environmental water testing
25
Dichromate/H2SO4
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • COD vario tube test
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH208-Contains Potassium dichromate. May produce an allergic reaction.
Precautionary Statement
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, disilver(1) sulphate, potassium dichromate