missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ DPD No. 4 Tablets

Convenient blister packaging allows user to push tablet through foil directly into sample

Brand:  Lovibond™ 511220BT

Product Code. 15371935

  • 25.75 € / Pack of 100
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Convenient blister packaging allows user to push tablet through foil directly into sample

  • Each tablet hermetically sealed in individual aluminum foil pocket that protects it from environmental conditions for longer shelf life
  • For use with MC500™ Multi-Parameter Handheld Colorimeter (Cat. No. 15-398-470) and SP600 Spectrophotometer (15-398-320)
  • Tests for active oxygen in water samples
  • Range: 0.1 to 10mg/L

Specifications

Specifications

White
6.2
Water analysis
0.1 to 10mg/L
DPD
Potassium Iodide 1 to 10%, N,N-diethylbenzene-1,4-diammonium Sulphate 0.1 to 1%, non-hazardous components >90%
Tablet
MC500 colorimeter and SP600 spectrophotometer
20°C
DPD No. 4 Tablet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.