missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Calibration Set for Infrared Lab Turbidimeter

For use with TB300-IR Lab Turbidimeter.

Brand:  Lovibond™ 194150


Product Code. 11779059

  • 234.56 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: Standards for 0.1, 20, 200, 800 NTU
This item is not returnable. View return policy

Description

Description

For use with TB300-IR Lab Turbidimeter.

For use with TB300-IR Lab Turbidimeter.
Specifications

Specifications

Calibration Kit
TB 300-IR Lab Turbidimeter
Standards for 0.1, 20, 200, 800 NTU
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • T-CAL Standards 800 NTU / 200 NTU / 20 NTU / < 0.1 NTU

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : methenamine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.