missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ NC45 Non Contact Benchtop Spectrophotometer

Allows sample measurements from a distance, eliminating surface distortion caused by contact based measurement methods

Brand:  Lovibond™ 400810

Product Code. 15554283

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

It is used for measuring many types of wet and dry samples including powders, pastes, gels, plastics and paints. 'Non-contact' design allows detailed color measurement of irregular shaped items such as granules and pellets.

  • Geometry: 45°/0°
  • Full system LED illumination
  • Line of Sight™ visibility to sample
  • Active Visual Targeting™ projects prominent illuminated target ring onto sample
  • Dual measurement spot sizes: 0.25 in. (6mm) and 0.5 in. (12mm)
  • Integrated gloss sensor provides 60° correlated gloss values
  • Flexible Quality Control software
  • Color scale: CIE 94, CIE L™ u™ v™ Color Space, CIE L™a™b, CIELAB (L™C™h), CMC Tolerancing, Whiteness Index (ASTM E 313), xyY chromaticity co-ordinates, XYZ tristimulus values, Yellowness Index (ASTM E 313), Delta E™ color difference, miscommunication of color data, spectral data (absorbance), a guide to path length by color scale

Specifications

Specifications

UL 61010-1, CSA 22.2 No. 1010.1 and IEC (EN) 61010-1
9.75 x 14 x 16 in. (24.8 x 35.6 x 40.6 cm)
400 to 700 nm
16 in.
9.75 in.
Double Beam
2.24 kg
35.6 cm
USB 2
24 V
Food and beverages, coatings, textile, chemicals, pharmaceutical and cosmetics, medical and clinical, household products, Plastics, sugar solutions, syrups and honey
40.6 cm
24.8 cm
4.931 lb.
14 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.