missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ PD250 Reagent Dispenser Refill: DPD Total Chlorine
For use with SC450 Chlorine and pH colorimeter, MC500 colorimeter and SP600 spectrophotometer
Brand: Lovibond™ 530150
This item is not returnable.
View return policy
Specifications
For chlorine testing | |
Includes 2 vial of DPD total chlorine reagent | |
Replacement Total Chlorine Reagent |
Used with SC450 chlorine and pH colorimeter, MC500 colorimeter and SP600 spectrophotometer | |
250 Tests |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 2 x 250 TESTS VARIO POWDER TOTAL CHLORINE 10 ML .
Signal Word
- Danger
Hazard Category
- Specific target organ toxicity Category 1
Hazard Statement
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P314-Get medical advice/attention if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : potassium iodide, EDTA
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction