Learn More
Lovibond™ Phenol Red Tablet
Lovibond Phenol Red Tablet, Orange, Composition: Sodium Chloride, Format: Tablet, For Use With: Water analysis, Colorimeter and spectrophotometer, Method: Phenol red, pH Range: 6.5 to 8.4, Shelf Life: 10 Years, Storage Requirements: 20 deg.C
Brand:  Lovibond™ 511790BT
Product Code. 15361965
  • 25.75 € / Pack of 100
Explore more special offers
Description
Description
Lovibond Phenol Red Tablet, Orange, Composition: Sodium Chloride, Format: Tablet, For Use With: Water analysis, Colorimeter and spectrophotometer, Method: Phenol red, pH Range: 6.5 to 8.4, Shelf Life: 10 Years, Storage Requirements: 20 deg.C
Specifications
Specifications
Orange
6.5 to 8.4
Water analysis
20°C
Phenol Red Tablet
Sodium Chloride
Tablet
Colorimeter and spectrophotometer
Phenol red
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lovibond™ Phenol Red Tablet > Phenol Red Tablet; Quantity: 100/Pk.