missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Powder Packs™ for Free Chlorine
Premeasured unit dose DPD reagents for water quality testing
Brand: Lovibond™ 530103
This item is not returnable.
View return policy
Description
Premeasured unit dose DPD reagents for water quality testing
- For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
- Also suitable for use in Hach™ instruments and test kits
- Easy-to-open, sealed foil packets eliminate risk of cross-contamination
- Available for 5, 10, and 25mL sample sizes
- Extended shelf life of up to 5 years
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted DPD method
Specifications
US EPA | |
Powder | |
SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer | |
0 to 2 mg/L | |
0.33 oz. |
10 mL | |
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
1000 Pack | |
DPD Free Chlorine Reagent |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
Hazard Statement
- EUH210-Safety data sheet available on request.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lovibond™ Powder Packs™ for Free Chlorine > Free Chlorine; 10mL; Pack of 1000
Spot an opportunity for improvement?Share a Content Correction