missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Powder Packs™ for Free Chlorine
Premeasured unit dose DPD reagents for water quality testing
Brand: Lovibond™ 530110
This item is not returnable.
View return policy
Description
Premeasured unit dose DPD reagents for water quality testing
- For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
- Also suitable for use in Hach™ instruments and test kits
- Easy-to-open, sealed foil packets eliminate risk of cross-contamination
- Available for 5, 10, and 25mL sample sizes
- Extended shelf life of up to 5 years
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted DPD method
Specifications
US EPA | |
Powder | |
SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer | |
0 to 5 mg/L | |
0.84 oz. |
25 mL | |
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
100 Pack | |
DPD Free Chlorine Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lovibond™ Powder Packs™ for Free Chlorine > Free Chlorine; 25mL; Pack of 100
Spot an opportunity for improvement?Share a Content Correction