Learn More
Lovibond™ Total Chlorine Powder Packs™
Premeasured unit dose DPD reagents for water quality testing
Brand:  Lovibond™ 530130
 View more versions of this product
Product Code. 15351655
  • 28.20 € / Pack of 100
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Premeasured unit dose DPD reagents for water quality testing
  • For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed foil packets eliminate risk of cross-contamination
  • Available for 5, 10, and 25mL sample sizes
  • Extended shelf life of up to 5 years
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted DPD method
Specifications
Specifications
US EPA
Powder
SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer
0 to 5 mg/L
0.84 oz.
25 mL
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
100 Pack
DPD Total Chlorine Reagent