Learn More
Lovibond™ RD125 Reactor
Used for the chemical digestion of water samples, such as in the determination of COD, TOC, Total Phosphate and Total Nitrogen
Brand:  Lovibond™ 2418940
Product Code. 12681735
  • 1105.00 € / Each
Explore more special offers
Description
Description
Used for the chemical digestion of water samples, such as in the determination of COD, TOC, Total Phosphate and Total Nitrogen
  • Required temperatures and reaction times are preprogrammed, and easy to select using the keypad
  • Users can pre-select reaction times of 30, 60 and 120 minutes or can set it to operate continuously
  • Power: 100-240 VAC, 50-60Hz
Specifications
Specifications
CE, UL, TUV
115/230 V
9.49 cm
10.83 in.
Round Metal block in Epoxy covered housing
7.9 lb.
6.1 in.
10.83 x 6.1 x 3.74 in. (27.51 x 15.49 x 9.49 cm)
3.74 in.
RD125 Reactor, 16mm COD vial adapter, instructions
27.51 cm
0 to 15000 mg/L
3.58 kg
15.49 cm