Learn More
Lovibond™ Ammonia Reagent Set, for Tube Testing, High Range
Premeasured unit dose reagent for water quality testing
Brand:  Lovibond™ 535650
Product Code. 15248103
  • 128.00 € / Each
Explore more special offers
Includes: Ammonia Cyanurate Reagent (5mL sample, 100 pack), Ammonia Salicylate Reagent (5mL , 100 pack), HR AM Dilution Tube (50 ct.)
Description
Description
Premeasured unit dose reagent for water quality testing
  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Specifications
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
5mL ammonia cyanurate reagent (100 pack), 5mL ammonia salicylate reagent (100 pack), 50 ct. HR AM dilution tube
0 to 50mg/L
High Range Ammonia Reagent Set
Hach™ and other manufacturers test kits, colorimeters and spectrophotometers
25
Salicylate
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • high range vario am test tube reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : lithium hydroxide monohydrate, sodium dichloroisocyanurate,sodium salicylate, sodium nitroprusside