Learn More
Lovibond™ Total Nitrogen Reagent Set, for Tube Testing, High Range
Premeasured unit dose reagent for water quality testing
Brand:  Lovibond™ 535560
Product Code. 15341695
  • 199.00 € / Pack of 50
Explore more special offers
Includes: Total N Persulfate Reagent, Total Nitrogen Reagent A, Total Nitrogen Reagent B, Total Nitrogen Hydroxide HR Tubes, Total Nitrogen Acid LR/HR Tubes, Deionized Water, 100mL
Description
Description
  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Specifications
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
Total N persulfate reagent (50 pack), total nitrogen reagent A (50 pack), total nitrogen reagent B (50 pack), 50 ct. total nitrogen hydroxide HR tubes, 50 ct. total nitrogen Acid LR/HR tubes, 100mL deionized water
0 to 150mg/L
High Range Total Nitrogen Reagent Set
Hach™ and other manufacturers test kits, colorimeters and spectrophotometers, reactor
25
Persulfate Digestion
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lovibond™ Total Nitrogen Reagent Set, for Tube Testing, High Range > Total Nitrogen set; High Range