missing translation for 'onlineSavingsMsg'
Learn More

SPEX™ Low Binding Silica Grinding Beads

SPEX™ Low Binding Silica Grinding Beads are acid washed silica grinding beads that are chemically treated to keep samples from binding to titer plate.

Brand:  SPEX™ 2167

 View more versions of this product

Product Code. 15300432

  • 371.00 € / 200g
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Used for lysing dilute samples of cells in the Geno/Grinder
  • Treated to remove fine particles and contaminates
  • Coated to reduce non-specific binding of nucleic acids and proteins
  • Bead size approximately 400 μm and 800 μm
  • Compatible with all SPEX™ SamplePrep Titer Plates and Vial Sets
Specifications

Specifications

400 μm beads
Silica
200 g bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers