Learn More
3M™ Particulate Respirator, Series C100
Lightweight and comfortable particulate respirators provide effective protection against dust and mist.
Brand:  3M™ 7000088841
 View more versions of this product
Product Code. 13507010
  • 4.06 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
Provides cost-effective, lightweight, comfortable, and effective protection against dust and mist.
  • The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes
  • Exhalation valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FFP1
General Purpose Particulate Respirator
C111
No
Polypropylene
Yes
Non-sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Particulate Respirator, Series C100 > Model: C111; Filter efficiency: FFP1; Valved