Learn More
3M™ Particulate Respirator, Series C100
Provide comfortable, effective respiratory protection against dust and mist. The 3M C100 Series Particulate Respirator is lightweight and comfortable.
Brand: 3M™ 7000088842
This item is not returnable.
View return policy
Description
Provides cost-effective, lightweight, comfortable, and effective protection against dust and mist.- The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes
- Exhalation valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
Specifications
FFP2 NR D | |
Polypropylene | |
Unvalved Particulate Respirator | |
C102 |