Learn More
BRAND™ Soda Lime Glass Culture Tubes with Screw Cap
Manufactured from soda lime glass. BRAND™ Soda Lime Glass Culture Tubes with Screw Cap include a polypropylene screw cap with white elastomer seal and GL thread. Autoclavable at 121°C.
Brand:  BRAND™ 113931
 View more versions of this product
Product Code. 10372941
  • 106.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from soda lime glass.
  • Includes polypropylene screw cap with white elastomer seal and GL thread.
  • Autoclavable (121°C).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
Soda Lime Glass
GL 14
Screw Cap
100 mm
100/Pk.
12 mm
Culture Tube