Learn More
Lenz Laborglasinstrumente™ Low-Form Weighing Bottles
For the precise and secure weighing of samples in the laboratory. Lenz Laborglasinstrumente™ Low-Form Weighing Bottles are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 05560516
 View more versions of this product
Product Code. 15320895
  • 227.00 € / Pack of 6
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With interchangeable NS stopper or cap
  • With fused-in size (mm) and capacity (mL)
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
45 mL
1 Pack
Weighing Bottle
Button Cover
Low
30 mm
60 mm