missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Low pl Marker Kit

The markers may be run in parallel with the experimental samples on isoelectric focusing gels.

Brand:  Cytiva 29443548

330.73 EUR valid until 2024-12-31
Use promo code "21982" to get your promotional price.


Product Code. 16592532

  • 404.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • pl markers are lyophilized mixtures of stable, salt-free, highly purified proteins
  • These markers are not prestained
  • Amylglucosidase - 3.50 pI (Native)
  • Methyl red (dye) - 3.75 pI (Native)
  • Glucose oxidase - 4.25 pI (Native)
  • Trypsin inhibitor - 4.55 pI (Native)
  • b-Lactoglobulin A - 5.20 pI (Native)
  • Carbonic anhydrase B (bovine) - 5.85 pI (Native)
  • Carbonic anhydrase B (human) - 6.55 pI (Native)
  • Not for use in denaturing (urea) systems
  • Store at 4°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
3.5 to 6.5
To be used for isoelectric focusing (IEF)
4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.