Learn More
Idex Low-Pressure PEEK Tees
Combine, split, or intersect fluid streams throughout your system
Brand:  Idex P-712
 View more versions of this product
Product Code. 13060390
  • 82.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
Use Tee connectors in a variety of low-pressure situations throughout your fluidic system. Handy mounting holes make it a snap to mount connectors. Tee connector assemblies come complete with nuts and ferrules.
Specifications
Specifications
Tees and Crosses
1000 psi
0.50 mm
0.020 in.
PEEK