missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Low Profile Tubes and Domed Caps

Low profile strip tubes for PCR and qPCR applications.

126.00 € - 265.00 €

Specifications

Color Natural
View More Specs

Products 3
Product Code Brand Color Closure Included Price Quantity & Availability  
Product Code Brand Color Closure Included Price Quantity & Availability  
10364703
View Documents Promotion Details
Thermo Scientific™
AB0775
Natural Dome Caps
265.00 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
11514804
View Documents Promotion Details
Thermo Scientific™
AB-0847
Natural No This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10776163
View Documents Promotion Details
Thermo Scientific™
AB0772
Natural Dome Caps
167.00 €
Pack of 120
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ideal for applications requiring low reaction volumes, Thermo Scientific™ Low Profile Strip Tubes are compatible with 0.2mL cycler blocks.

The low profile design of the Thermo Scientific Low Profile Strip Tubes reduces the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Thermo Scientific low profile products are especially recommended for use with reaction volumes below 20 μL.

The low profile design of the Thermo Scientific strip tubes without caps reduces the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Low profile products are especially recommended for use with reaction volumes below 20 μL.

Highlights

  • Suitable for 0.2 mL thermal cycler blocks
  • 150 μL maximum tube capacity when capped
  • Low profile to reduce dead space and increase PCR efficiency
  • Labelled A-H end tabs (8-strip) and 1-12 end tabs (12-strip)
Applications
  • PCR
  • qPCR/Real-Time PCR

Hazardous: No
Storage Condition: Ambient

PCR, qPCR and Real-Time PCR.

Certifications

Certified free from human DNA, RNases, and DNases.

Specifications

Specifications

Natural
Videos
SDS
Documents

Documents

Product Certifications