missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Low Serum Growth Supplement Kit (LSGS Kit)
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
The LSGS Kit provides, in separate vials, all the components of complete LSGS (S-003-10): fetal bovine serum, hydrocortisone, human epidermal growth factor, and basic fibroblast growth factor/heparin (stabilized with BSA). A vial of gentamycin/amphotericin solution (GA) is also included. Use of GA is optional. When a 500 ml bottle of medium is supplemented with LSGS or the components of the LSGS Kit, the final concentrations of the components in the supplemented medium are:
fetal bovine serum, 2% v/v
hydrocortisone (1μg/ml)
human epidermal growth factor, 10 ng/ml
basic fibroblast growth factor, 3 ng/ml
heparin, 10μg/ml
Specifications
Specifications
| Content And Storage | minus 20 degrees C |
| Cell Type | Epithelial Cells (Large Vessel), Fibroblast Cells (Dermal) |
| Product Type | Growth Supplement |
| Sterility | Sterile |
| Product Line | Cascade Biologics |
| Quantity | 1 Kit |
| Shipping Condition | Dry Ice |
| Species | Human |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction