missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Low Speed Benchtop Centrifuge

LMC-3000 Low speed benchtop centrifuge with interchangeable rotors for accommodating centrifuge tubes (from 2 to 50ml) or microplates.

Brand:  Grant Instruments™ LMC-3000

1854.95 EUR valid until 2025-12-31
Use promo code "24282" to get your promotional price.


Product Code. 11461048

  • 1985.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

LMC-3000 Low speed benchtop centrifuge with interchangeable rotors for accommodating centrifuge tubes (from 2 to 50ml) or microplates.

The unit detects rotor imbalance for reduced wear and decreased noise and vibration, and comes with a 1 to 90 minutes timer with automatic turn-off when a timed procedure completes. Note that this unit is non-refrigerated and does not have temperature control features.
  • Interchangeable rotors for accommodating centrifuge tubes (10-15mL or 50mL) or microplates
  • Extremely quiet motor with 'soft' start up and run-down function to avoid jolting of samples
  • Compact design with small footprint
  • Auto 'IMBALANCE' stop and warning diagnostics
  • Two-year warranty Suitable for a wide range of analytical applications including biomedical, bio-organic and immunoenzyme analysis.
Specifications

Specifications

1,700 x g
No
Laxco Routine Clinical Microscope
2,000 rpm
4°C to 40°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.