Learn More
Low Temperature Viscosity Standard, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified low temperature viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.
Brand:  Paragon Scientific N1400B
 View more versions of this product
Product Code. 15578874
  • 98.00 € / 500mL
Explore more special offers
Description
Description
  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Standard Pack Size 500mL
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
  • Specifications
    Specifications
    N1400B
    500 mL
    172195 mPa.s (-13°C)
    Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Nominal values quoted