missing translation for 'onlineSavingsMsg'
Learn More

Low Temperature Viscosity Standard, Paragon Scientific™

Paragon ISO 17025 / ISO 17034 certified low temperature viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.

Brand:  Paragon Scientific JF1-L


 View more versions of this product

Product Code. 15781928

  • 98.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Standard Pack Size 500mL
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
  • Specifications

    Specifications

    JF1-L
    500 mL
    5989.0 mPa.s (-40°C)
    Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
    Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling

    Safety and Handling

    Signal Word
    • Danger

    Hazard Category
    • Flammable liquid Category 3
    • Skin corrosion/irritation Category 2
    • Specific target organ toxicity Category 3
    • Aspiration toxicity Category 1
    • LONG-TERM AQUATIC HAZARD Chronic 2

    Hazard Statement
    • H226-Flammable liquid and vapour.
    • H304-May be fatal if swallowed and enters airways.
    • H315-Causes skin irritation.
    • H336-May cause drowsiness or dizziness.
    • H411-Toxic to aquatic life with long lasting effects.

    Precautionary Statement
    • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
    • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
    • P331-Do NOT induce vomiting.
    • P370+P378-In case of fire: Use to extinguish.

    Supplemental information
    • MIXTURE LIST-Contains : kerosine

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Nominal values quoted