Learn More
Novus Biologicals™ LPS from E. Coli, TLR4 ligand
Applications: Functional
Brand:  Novus Biologicals™ NBP2-25295
 View more versions of this product
Product Code. 18604030
  • 790.00 € / 10mg
Explore more special offers
Description
Description
Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs). Use in functional reported in scientific literature (PMID 26121241).
Lipopolysaccharides from Escherichia coli 0111:B4 Gamma-irradiated, BioXtra, suitable for cell culture Dilution: Functional Purity: for ion exchange chromatography Reconstitute with 1 ml sterile balanced salt solution or tissue culture medium to the vial (1 mg) and gently swirl until the powder dissolves. Reconstituted product may be further diluted to desired working concentrations.
Specifications
Specifications
10 mg
LPS from E. Coli, TLR4 Ligand