Learn More
IKA™ LR 1000.3 Laboratory Reactor Vessel
Shop All IKA Products
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Each
Description
- The ultimate systems when it comes to optimizing and reproducing chemical reaction, mixing, dispersing, and homogenization processes on a laboratory scale.
- Can be individually customized to perform numerous tasks, including: production of creams and lotions, and the grinding and disintegrating of solids and fibers in liquids or polymers.
- Single-walled or double-walled vessels made from borosilicate glass or stainless steel are available, each with or without bottom discharge valve.
- Functionality, safety, and longevity are the main goals in the development of IKA laboratory reactors.
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Material | FFKM, PEEK, Borosilicate Glass (3.3) |
| Includes | 6 Connectors |
| Product Type | Reactor vessel |
| Standard Taper Joint | 2 x NS 29, 2 x NS 14, 2 x GL 14 |