missing translation for 'onlineSavingsMsg'
Learn More

IKA™ LR 1000 Basic System

The LR 1000 basic is a cost efficient, modular laboratory reactor designed for optimizing chemical reaction processes as well as for mixing, dispersing and homogenization applications at the laboratory scale.

Brand:  IKA™ 0020119960

Product Code. 17832804

  • 7951.00 € / Each
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The LR 1000 basic is a cost efficient, modular laboratory reactor designed for optimizing chemical reaction processes as well as for mixing, dispersing and homogenization application at the laboratory scale.
  • The reactor is especially designed for the unique application requirements that define the cosmetic and pharmaceutical industries.
  • The temperature of the medium can be as hight as 120°C.
  • The temperature of the heating source, which is located at the bottom of hte vessel is controlled accordingly to reach the set temperature inside the medium. A PT 100.30 temperature sensor and the corresponding receptacle LR 1000.61 for the lid are included with this package.
Specifications

Specifications

1000 mL
22 kg
Stainless Steel (316L)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.