missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ LSE™ Fixed-Angle Rotors
Rotors with different tube size and capacity configurations
Brand: Corning™ 480137
603.05 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
This is an optional rotor for 6 x 50 mL tubes, for use with Corning™ LSETM Compact Centrifuge.Specifications
Rotor | |
6,000 rpm | |
Fixed |
12 x 15 mL | |
Corning™ LSE™ Compact Centrifuge |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction