missing translation for 'onlineSavingsMsg'
Learn More

Corning™ LSE™ Nutating Mixer, GB Plug

A small, personal-sized 3D mixer that is ideal for ambient and cold room experiments. Ideal for applications such as immunohistochemistry, mixing and resuspending small containers in blood or centrifuge tubes securely held to the mixer with a dimpled mat.

Brand:  Corning™ 6722

Product Code. 17710799

  • 333.26 € / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Small footprint
  • Cold room compatible
  • Dimpled Mat
  • 3D Motion
  • Ideal for mixing and resuspending samples
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.8 kg
120 V
0.03 A
cETLus
50/60 Hz
14.6 cm
≤80%, Non-condensing
2 Years
20°
4°C to 40°C
1.75 lb.
GB Plug
5.75 in.
24 x 17 cm (9.5 x 6.75 in.)
Dimpled Mat
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.