missing translation for 'onlineSavingsMsg'
Learn More

LUF 5834, Tocris Bioscience™

Product Code. 30002063
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent adenosine A2A and A2B receptor partial agonist

LUF 5834 is a potent A2A and A2B adenosine receptor partial agonist (Ki = 2.6 nM and EC50 = 12 nM respectively). Also exhibits selectivity for A1 over A3 (Ki values are 2.6 and 538 nM respectively).

Chemical Identifiers

CAS 333962-91-7
Molecular Formula C17H12N6OS
Molecular Weight (g/mol) 348.384
InChI Key LOLJUQOCQBYBGT-UHFFFAOYSA-N
Synonym 2-1h-imidazol-2-yl methyl thio-6-amino-4-4-hydroxyphenyl pyridine-3,5-dicarbonitrile, 2-amino-4-4-hydroxyphenyl-6-1h-imidazol-2-ylmethyl thio-3,5-pyridinecarbonitrile, 2-amino-4-4-hydroxy-phenyl-6-1h-imidazol-2-ylmethylsulfanyl-pyridine-3,5-dicarbonitrile
PubChem CID 10066330
IUPAC Name 2-amino-6-(1H-imidazol-2-ylmethylsulfanyl)-4-(4-oxocyclohexa-2,5-dien-1-ylidene)-1H-pyridine-3,5-dicarbonitrile
SMILES C1=CC(=O)C=CC1=C2C(=C(NC(=C2C#N)SCC3=NC=CN3)N)C#N

Specifications

Quantity 10 mg
Formula Weight 348.38
Chemical Name or Material LUF 5834
missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'