missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luff-Schoorl Reagent, Honeywell Fluka™
Volumetric, 25 g/L
64.60 € - 264.36 €
Specifications
Chemical Name or Material | Luff-Schoorl Reagent |
---|---|
CAS | 497-19-8, 7732-18-5, 7758-98-7 |
UN Number | UN3082 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15624170
|
Honeywell-Fluka
34400-1L |
1 L |
64.60 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15634170
|
Honeywell-Fluka
34400-5L-VP |
5 L |
264.36 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Luff-Schoorl Reagent | |
497-19-8, 7732-18-5, 7758-98-7 | |
UN3082 |
Safety and Handling
P273-P280-P305 + P351 + P338