Learn More
Pro-Lab Diagnostics™ Lugol’s Iodine Stain
Use for convenient Gram staining to easily identify gram positive or negative organisms. Pro-Lab Diagnostics™ Lugol’s Iodine Stain is a ready-to-use microbiological stain manufactured to the highest standards, according to all current regulatory requirements, and CE labelled.
Brand:  Pro-Lab Diagnostics™ PL.7052
Product Code. 12801823
  • 70.00 € / 500mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Pro-Lab Diagnostics™ stains are manufactured in purpose-designed laboratories to the highest standards using the finest raw materials. Strict quality control and filtering techniques are employed prior to release to ensure that all stains offer excellent performance characteristics.
  • Ready-to-use stain in convenient and economical sizes
  • Apply after staining with crystal violet, but before ethanol
  • Stains the peptidoglycan of gram positive organisms
Specifications
Specifications
Reagent
500mL
Liquid
Dark brown
Store in a cool, well-ventilated place
Lugol's Iodine
(EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark
Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria
Ethanol 2.5 – <5%, Potassium iodide 1 – <2.5%, Iodine 0.5 – <1%, Methanol 0.25 – <0.5%
Gram Stain