Learn More
Emma Safety Footwear Lukas Safety Shoes
Includes a steel toe cap, a polyurethane outer toe cap and an easy-twist sole that provides a sticky grip.  Available in a variety of widths.
Brand:  Emma Safety Footwear MM73054835
 View more versions of this product
Product Code. 15958931
  • 127.00 € / 1 pair
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather
  • GripForce™ Easy Twist
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011
Black
Hydro-Tec™ Sanitized Silver
35
Laces
Lukas
Leather
Steel