Learn More
Thermo Scientific™ Accessories for Luminoskan™ Ascent Microplate Luminometer
Run a variety of luminometric research applications with this compact, robust instrument certified as DLR-Ready.
Brand:  Thermo Scientific™ 2805621
 View more versions of this product
Product Code. 12413458
  • 10500.00 € / Each
Explore more special offers
Description
Description
Run a variety of luminometric research applications with the Thermo Scientific™ Luminoskan™ Ascent Microplate Luminometer, a compact and robust instrument offering versatile plate formats, fast reading speeds and up to three dispensers. This instrument has passed the DLReady™ validation at Promega Corporation™ and is certified as DLReady. The Luminoskan Ascent Microplate Luminometer is validated for Thermo Scientific™ Pierce™ Luciferase Assays.
Fast reading speed:
  • Reads a 96-well plate in just 15 seconds
  • Essential for kinetic applications, such as enzyme kinetics and phagocytosis assays
Onboard dispensers for exact follow-up of kinetic reactions:
  • Equipped with up to three reagent dispensers for fast kinetic assays, such as ATP assays
  • Supports simultaneous dispensing and reading, enablingmonitoring of fast kineticmeasurements from the very start of the reaction
  • Offers very low dead volume and backflush capability for conservation of expensive reagents
High sensitivity:
  • DLReady certified for excellent sensitivity
  • Fiberless direct illumination optics ensuresmaximal light collection
  • High sensitivity, a wide dynamic range and low crosstalk for accurate and precise results in luminometric assays
Specially designed for automation:
  • Reads up to 384-well plates for increased assay throughput
  • Easily integrates with automated systems
Includes:
PC Software
Recommended for:
  • Reporter gene assays
  • Immunoassays with luminescent substrates
  • Cell proliferation and cytotoxicity assays
  • Intracellular Ca2+ assays
  • ATP assays
  • Phagocytosis assays
  • Reactive oxygen assays
  • microbiological assays
  • Enzyme assays
  • BRET assays
  • ADmEtox
Services
1 year
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1st Dispenser Kit for Luminoskan
384-well Plate
PC Software
Robot
Luminoskan