missing translation for 'onlineSavingsMsg'
Learn More
Learn More
American Educational Products Lung/Breath Volume Kits
Investigate pulmonary fitness.
49.94 € - 81.81 €
Specifications
Grade | 3 to 9 |
---|---|
For Use With (Application) | To measure lung volume with reusable components |
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
13152191
|
American Educational Products
6050 |
Lung Volume Kit | Durable plastic bag calibrated to 6l, mouthpiece and mouthpiece holder | N/A | |||||
13142191
|
American Educational Products
6109 |
Breath Volume Kit | Durable plastic bag, mouthpiece and complete instruction guide | N/A | |||||
Description
- Lung Volume Kit allows you to measure lung volume with reusable components
- Breath Volume Kit measures tidal and respiratory minute volume
Specifications
3 to 9 | |
To measure lung volume with reusable components |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title