Learn More
Novus Biologicals™ Lung Tissue Slides (Bronchioloalveolar Carcinoma)- Paraffin
Brand:  Novus Biologicals™ NBP2-30282
Product Code. 18132135
  • 207.00 € / Pack of 5
Explore more special offers
Description
Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Specifications
Specifications
Human
Immunohistochemistry-Paraffin
Human
Lung Tissue Slides (Bronchioloalveolar Carcinoma)
This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Bronchioloalveolar Carcinoma