missing translation for 'onlineSavingsMsg'
Learn More
Lutetium foil, 0.5mm (0.02in) thick, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 15440987

Lutetium foil, 0.5mm (0.02in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 15440987
25 x 25 mm, Mylar under Argon
Click to view available options
Quantity:
25 x 25 mm
Packaging:
Mylar under Argon
This item is not returnable. View return policy

Product Code. 15440987

Brand: Thermo Scientific Alfa Aesar 010229.FF

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

Lutetium is used as an additive for special steel, non-ferric alloy, hydrogen storage alloy, also reducer for making other rare Earth metal. Pipe materials of reactor system. Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Specifications

CAS 7439-94-3
Molecular Formula Lu
MDL Number MFCD00011098
Quantity 25 x 25 mm
Synonym lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
Solubility Information Insoluble in water.
InChI Key OHSVLFRHMCKCQY-UHFFFAOYSA-N
SMILES [Lu]
IUPAC Name lutetium
Molecular Weight (g/mol) 174.97
PubChem CID 23929
ChEBI CHEBI:33382
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈3.1g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Lutetium foil, 0.5mm (0.02 in.) thick
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lutetium foil, 0.5mm (0.02in) thick, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.