missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) 2,4-pentanedionate, REacton™, 99.9% (REO)

Product Code. p-7049196
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11322288

Brand: Thermo Scientific Alfa Aesar 013189.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17966-84-6 | C15H21LuO6 | 472.29 g/mol

Lutetium(III) 2,4-pentanedionate is involved in refluxing 1,2,4-trichlorobenzene. It is also used as catalyst of organic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17966-84-6
Molecular Formula C15H21LuO6
Molecular Weight (g/mol) 472.29
MDL Number MFCD00078030
InChI Key HXFHOORFYPCGNF-UHFFFAOYSA-N
Synonym lutetium iii acetylacetonate hydrate, lutetium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy lutetio oxy pent-3-en-2-one
PubChem CID 24282830
SMILES [Lu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 1 g
Solubility Information Insoluble in water.
IUPAC Name lutetium(3+) tris(2,4-dioxopentan-3-ide)
Formula Weight 472.3
Percent Purity 99.9%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lutetium(III) 2,4-pentanedionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.