missing translation for 'onlineSavingsMsg'
Learn More
Lutetium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11352518

Lutetium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11352518
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11352518

Brand: Thermo Scientific Alfa Aesar 014571.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207500-05-8
Molecular Formula C6H9LuO6
Molecular Weight (g/mol) 352.10
MDL Number MFCD00211310
InChI Key YMPHWTSLMNFNFB-UHFFFAOYSA-K
IUPAC Name lutetium(3+) triacetate
SMILES [Lu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 352.11 (Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lutetium(III) acetate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lutetium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.