missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific™

Product Code. 10786373
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-66-8
Molecular Formula Cl3Lu
Molecular Weight (g/mol) 281.33
MDL Number MFCD00011099

Specifications

Melting Point 905°C
Color White to Gray
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.9%
Linear Formula LuCl3
Solubility Information Solubility in water: soluble
Formula Weight 281.33
Percent Purity 99.90%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Lutetium(III) chloride
Show More Show Less
compliance-icons
Product Identifier
  • Lutetium(III) chloride, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.